O-Tritylhydroxylamine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

T3860-1G T3860-5G
TCT3860-1GEA 50 USD
TCT3860-1G TCT3860-5G
O-Tritylhydroxylamine ≥98.0% (by HPLC, total nitrogen)
O-Tritylhydroxylamine
Formula: C₁₉H₁₇NO
MDL Number: MFCD00042818
CAS Number: 31938-11-1
EINECS: 250-868-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 84.0 to 88.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR