Etofibrate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-[[2-(4-Chlorophenoxy)-2-methylpropanoyl]oxy]ethyl Nicotinate, Nicotinic acid 2-[[2-(4-Chlorophenoxy)-2-methylpropanoyl]oxy]ethyl ester, 2-((2-(4-Chlorophenoxy)-2-methylpropanoyl)oxy)ethyl nicotinate, 3-Pyridinecarboxylic acid 2-[2-(4-Chlorophenoxy)-2-methyl-1-oxopropoxy]ethyl ester, 2-[2-(4-Chlorophenoxy)-2-methyl-1-oxopropoxy]ethyl 3-Pyridinecarboxylate

E1359-250MG E1359-50MG
TCE1359-250MGEA 495 USD
TCE1359-250MG TCE1359-50MG
Etofibrate ≥98.0% (by GC)
Etofibrate
Formula: C₁₈H₁₈ClNO₅
MW: 363.79 g/mol
Boiling Pt: 214 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00865727
CAS Number: 31637-97-5
EINECS: 250-743-1
Merck Index: 14,03877

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR