2,4,6-Tris(4-butoxy-2-hydroxyphenyl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
T3870-1G
T3870-5G
TCT3870-1GEA
95
USD
TCT3870-1G
TCT3870-5G
2,4,6-Tris(4-butoxy-2-hydroxyphenyl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2,4,6-Tris(4-butoxy-2-hydroxyphenyl)-1,3,5-triazine
Formula:
C₃₃H₃₉N₃O₆ |
CAS Number:
3135-19-1 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...