3,4-Dihydro-5-hydroxy-1H-quinolin-2-one ≥98.0% (by total nitrogen basis)

Supplier: TCI America

D5669-1G D5669-5G
TCD5669-1GEA 68 USD
TCD5669-1G TCD5669-5G
3,4-Dihydro-5-hydroxy-1H-quinolin-2-one ≥98.0% (by total nitrogen basis)
3,4-Dihydro-5-hydroxy-1H-quinolin-2-one
Formula: C₉H₉NO₂
MDL Number: MFCD01862194
CAS Number: 30389-33-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 239.0 to 243.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR