2-(2-Hydroxyphenyl)-1H-benzimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H1736-25G H1736-5G
TCH1736-25GEA 393 USD
TCH1736-25G TCH1736-5G
2-(2-Hydroxyphenyl)-1H-benzimidazole ≥98.0% (by GC, titration analysis)
2-(2-Hydroxyphenyl)-1H-benzimidazole
Formula: C₁₃H₁₀N₂O
MDL Number: MFCD00022668
CAS Number: 2963-66-8

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 242.0 to 246.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR