Mono-2-(methacryloyloxy)ethyl Phthalate ≥90.0% (by titrimetric analysis) stabilised

Supplier: TCI America
Warning

Synonyms: Phthalic acid Mono-2-(methacryloyloxy)ethyl ester, 2-[[2-(Methacryloyloxy)ethoxy]carbonyl]benzoic acid, Mono-2-(methacryloyloxy)ethyl Hydrogen Phthalate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M2940-25G M2940-500G
TCM2940-25GEA 36 USD
TCM2940-25G TCM2940-500G
Mono-2-(methacryloyloxy)ethyl Phthalate ≥90.0% (by titrimetric analysis) stabilised
Mono-2-(methacryloyloxy)ethyl Phthalate
Formula: C₁₄H₁₄O₆
MW: 278.26 g/mol
MDL Number: MFCD00066298
CAS Number: 27697-00-3
EINECS: 248-616-0

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 85.0 %
Purity (Neutralization titration) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR