1-Ethylcyclohexyl methacrylate ≥98.0% (by GC) stabilized

Supplier: TCI America

Synonyms: Methacrylic acid 1-Ethylcyclohexyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

E1407-1G E1407-5G
TCE1407-1GEA 58 USD
TCE1407-1G TCE1407-5G
1-Ethylcyclohexyl methacrylate ≥98.0% (by GC) stabilized
1-Ethylcyclohexyl methacrylate
Formula: C₁₂H₂₀O₂
MW: 196.29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30470335
CAS Number: 274248-09-8

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR