Isopropoxybenzene ≥98.0% (by GC)

Supplier: TCI America

I1073-25G I1073-5G
TCI1073-25GEA 114 USD
TCI1073-25G TCI1073-5G
Isopropoxybenzene ≥98.0% (by GC)
Isopropoxybenzene
Formula: C₉H₁₂O
Boiling Pt: 176 °C
Melting Pt: −33 °C
MDL Number: MFCD00048288
CAS Number: 2741-16-4
EINECS: 220-370-9

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR