1-Ethylcyclopentyl methacrylate ≥98.0% (by GC) stabilised

Supplier: TCI America

Synonyms: Methacrylic acid 1-Ethylcyclopentyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

E1388-25G E1388-5G
TCE1388-25GEA 602 USD
TCE1388-25G TCE1388-5G
1-Ethylcyclopentyl methacrylate ≥98.0% (by GC) stabilised
1-Ethylcyclopentyl methacrylate
Formula: C₁₁H₁₈O₂
MW: 182.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23703079
CAS Number: 266308-58-1

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR