1,3-Dimethyl-1,3-diphenyl-1,3-divinyldisiloxane ≥98.0% (by GC)

Supplier: TCI America
Warning

D5557-25G D5557-5G
TCD5557-25GEA 177 USD
TCD5557-25G TCD5557-5G
1,3-Dimethyl-1,3-diphenyl-1,3-divinyldisiloxane ≥98.0% (by GC)
1,3-Dimethyl-1,3-diphenyl-1,3-divinyldisiloxane
Formula: C₁₈H₂₂OSi₂
Boiling Pt: 139 °C
MDL Number: MFCD00053860
CAS Number: 2627-97-6
EINECS: 220-101-5

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR