Dodecylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

D5537-1G D5537-5G
TCD5537-1GEA 42 USD
TCD5537-1G TCD5537-5G
Dodecylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
Dodecylamine hydrobromide
Formula: C₁₂H₂₇N·HBr
Melting Pt: 190 °C
MDL Number: MFCD30543762
CAS Number: 26204-55-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
water max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR