2-Oxohexahydro-2H-3,5-methanocyclopenta[b]furan-6-yl methacrylate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Methacrylic acid 2-Oxohexahydro-2H-3,5-methanocyclopenta[b]furan-6-yl ester, 5-Methacroyloxy-2,6-norbornane Carbolactone

O0536-25G O0536-5G
TCO0536-25GEA 430 USD
TCO0536-25G TCO0536-5G
2-Oxohexahydro-2H-3,5-methanocyclopenta[b]furan-6-yl methacrylate ≥98.0% (by GC)
2-Oxohexahydro-2H-3,5-methanocyclopenta[b]furan-6-yl methacrylate
Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
MDL Number: MFCD31715034
CAS Number: 254900-07-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 100.0 to 104.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR