1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethanamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

E1381-1G E1381-5G
TCE1381-1GEA 38 USD
TCE1381-1G TCE1381-5G
1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethanamine ≥98.0% (by GC, titration analysis)
1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethanamine
Formula: C₁₂H₁₉NO₄S
MW: 273.35 g/mol
MDL Number: MFCD27665207
CAS Number: 253168-94-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 119.0 to 123.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR