(3-Hydroxyphenyl)acetonitrile ≥95.0% (by GC)

Supplier: TCI America
Warning

H1615-1G H1615-5G
TCH1615-1GEA 112 USD
TCH1615-1G TCH1615-5G
(3-Hydroxyphenyl)acetonitrile ≥95.0% (by GC)
(3-Hydroxyphenyl)acetonitrile
Formula: C₈H₇NO
Boiling Pt: 106 °C
MDL Number: MFCD03265300
CAS Number: 25263-44-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR