2-(4-Methoxyphenyl)-1,3-dithiane ≥98.0% (by GC)

Supplier: TCI America

M3278-1G M3278-5G
TCM3278-1GEA 52 USD
TCM3278-1G TCM3278-5G
2-(4-Methoxyphenyl)-1,3-dithiane ≥98.0% (by GC)
2-(4-Methoxyphenyl)-1,3-dithiane
Formula: C₁₁H₁₄OS₂
MDL Number: MFCD01729781
CAS Number: 24588-72-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR