2,6-Diethyl-4-methylaniline ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D5517-1G
D5517-5G
TCD5517-1GEA
28
USD
TCD5517-1G
TCD5517-5G
2,6-Diethyl-4-methylaniline ≥98.0% (by GC)
2,6-Diethyl-4-methylaniline
Formula:
C₁₁H₁₇N Boiling Pt: 121 °C |
MDL Number:
MFCD00035950 CAS Number: 24544-08-9 EINECS: 246-307-5 |
Specification Test Results
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...