N,N-Dimethylethylenediamine dihydroiodide ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Warning

Synonyms: N,N-Dimethylethylenediammonium Diiodide, N1,N1-Dimethylethane-1,2-diamine dihydroiodide, 2-(Dimethylamino)ethylamine dihydroiodide

D5616-1G D5616-5G
TCD5616-1GEA 49 USD
TCD5616-1G TCD5616-5G
N,N-Dimethylethylenediamine dihydroiodide ≥98.0% (by total nitrogen basis)
N,N-Dimethylethylenediamine dihydroiodide
Formula: C₄H₁₂N₂·₂HI
MW: 343.98 g/mol
CAS Number: 244234-52-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR