Sodium Pyridine-2-sulfinate ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Sodium pyridine-2-sulphinate, Pyridine-2-sulphinic acid sodium salt

O0533-1G O0533-250MG
TCO0533-1GEA 311 USD
TCO0533-1G TCO0533-250MG
Sodium Pyridine-2-sulfinate ≥95.0% (by titrimetric analysis)
Sodium Pyridine-2-sulfinate
Formula: C₅H₄NNaO₂S
MW: 165.14 g/mol
MDL Number: MFCD20483664
CAS Number: 24367-66-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 93.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 283.0 to 287.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR