4-(1-Hydroxyethyl)phenol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Hydroxy-alpha-methylbenzyl alcohol

H1694-1G H1694-200MG
TCH1694-1GEA 189 USD
TCH1694-1G TCH1694-200MG
4-(1-Hydroxyethyl)phenol ≥98.0% (by GC)
4-(1-Hydroxyethyl)phenol
Formula: C₈H₁₀O₂
Boiling Pt: 118 °C
MDL Number: MFCD00016858
CAS Number: 2380-91-8
EINECS: 219-176-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 126.0 to 130.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR