N,N'-(Hexane-1,6-diyl)bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanamide] ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

H1704-25G H1704-5G
TCH1704-25GEA 54 USD
TCH1704-25G TCH1704-5G
N,N'-(Hexane-1,6-diyl)bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanamide] ≥98.0% (by HPLC, total nitrogen)
N,N'-(Hexane-1,6-diyl)bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanamide]
Formula: C₄₀H₆₄N₂O₄
MDL Number: MFCD00134697
CAS Number: 23128-74-7
EINECS: 245-442-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 158.0 to 162.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR