Dihydrorhodamine 6G

Supplier: TCI America

Synonyms: 2-[3,6-Bis(ethylamino)-2,7-dimethyl-9H-xanthen-9-yl]benzoic acid Ethyl ester, Ethyl 2-[3,6-Bis(ethylamino)-2,7-dimethyl-9H-xanthen-9-yl]benzoate

D5489-100MG D5489-25MG
TCD5489-100MGEA 52.17 USD
TCD5489-100MG TCD5489-25MG
Dihydrorhodamine 6G
Dihydrorhodamine 6G
Formula: C₂₈H₃₂N₂O₃
Storage Temperature: Refrigerator
CAS Number: 217176-83-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 6.0 to 6.6 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR