1-Ethyl-1H-pyrrole-2-carboxaldehyde ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Ethyl-1H-pyrrole-2-carbaldehyde, 2-Formyl-1-ethylpyrrole

E1416-1G E1416-5G
TCE1416-1GEA 87 USD
TCE1416-1G TCE1416-5G
1-Ethyl-1H-pyrrole-2-carboxaldehyde ≥98.0% (by GC)
1-Ethyl-1H-pyrrole-2-carboxaldehyde
Formula: C₇H₉NO
MW: 123.16 g/mol
Boiling Pt: 75 °C
MDL Number: MFCD11909543
CAS Number: 2167-14-8

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR