cvxIAA ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 5-(3-Methoxyphenyl)indole-3-acetic acid, 5-(3-MeOPh)-IAA, convex IAA

M3141-10MG M3141-50MG
TCM3141-10MGEA 137 USD
TCM3141-10MG TCM3141-50MG
cvxIAA ≥98.0% (by HPLC)
cvxIAA
Formula: C₁₇H₁₅NO₃
MW: 281.31 g/mol
CAS Number: 2127037-35-6

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR