2,2'-Oxydiacetyl chloride ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: Diglycolyl chloride, 2,2-Oxydiacetylchloride

O0459-25G O0459-5G
TCO0459-25GEA 175 USD
TCO0459-25G TCO0459-5G
2,2'-Oxydiacetyl chloride ≥97.0% (by GC, titration analysis)
2,2'-Oxydiacetyl chloride
Formula: C₄H₄Cl₂O₃
MW: 170.97 g/mol
Boiling Pt: 57 °C
MDL Number: MFCD00134471
CAS Number: 21062-20-4
EINECS: 244-186-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Colourless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR