2-Methyldecahydroquinoline (mixture of isomers) ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

M3189-1G M3189-5G
TCM3189-1GEA 165 USD
TCM3189-1G TCM3189-5G
2-Methyldecahydroquinoline (mixture of isomers) ≥98.0% (by titrimetric analysis)
2-Methyldecahydroquinoline (mixture of isomers)
Formula: C₁₀H₁₉N
Boiling Pt: 206 °C
MDL Number: MFCD20547663
CAS Number: 20717-43-5

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 % (total of isomer)
Concentration (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR