Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Ethyl 2,2-difluoro-2-(trimethylsilyl)acetate, 2,2-Difluoro-2-(trimethylsilyl)acetic acid Ethyl ester

E1314-1G E1314-5G
TCE1314-1GEA 90 USD
TCE1314-1G TCE1314-5G
Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate ≥97.0% (by GC)
Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate
Formula: C₇H₁₄F₂O₂Si
Boiling Pt: 55 °C
MDL Number: MFCD04973092
CAS Number: 205865-67-4
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid to cloudy liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR