3-Iodobiphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Iodo-biphenyl

I1055-1G I1055-5G
TCI1055-1GEA 43 USD
TCI1055-1G TCI1055-5G
3-Iodobiphenyl ≥98.0% (by GC)
3-Iodobiphenyl
Formula: C₁₂H₉I
MW: 280.11 g/mol
Boiling Pt: 178 °C
MDL Number: MFCD09032441
CAS Number: 20442-79-9

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR