Trimethyl(nonafluorobutyl)silane ≥98.0% (by GC)

Supplier: TCI America
Danger

T3594-1G T3594-5G
TCT3594-1GEA 104 USD
TCT3594-1G TCT3594-5G
Trimethyl(nonafluorobutyl)silane ≥98.0% (by GC)
Trimethyl(nonafluorobutyl)silane
Formula: C₇H₉F₉Si
Boiling Pt: 100 °C
MDL Number: MFCD20489405
CAS Number: 204316-01-8
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR