2-Phenylpropan-2-yl Benzodithioate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2-Phenylpropan-2-yl benzodithioate

P2638-1G P2638-5G
TCP2638-1GEA 226 USD
TCP2638-1G TCP2638-5G
2-Phenylpropan-2-yl Benzodithioate ≥98.0% (by HPLC)
2-Phenylpropan-2-yl Benzodithioate
Formula: C₁₆H₁₆S₂
MW: 272.42 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18252676
CAS Number: 201611-77-0

Order Now

Specification Test Results

Appearance Orange to Brown to Dark red powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 35.0 to 39.0 deg-C
Elemental analysis (Carbon) 69.1 to 72.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR