Gadobutrol monohydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

G0503-100MG G0503-25MG
TCG0503-100MGEA 160 USD
TCG0503-100MG TCG0503-25MG
Gadobutrol monohydrate ≥98.0% (by titrimetric analysis)
Gadobutrol monohydrate
Formula: C₁₈H₃₁GdN₄O₉·H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD29917846
CAS Number: 198637-52-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Elemental analysis (Nitrogen) 8.70 to 9.20 %
Water 2.6 to 3.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR