2-Oxotetrahydrofuran-3-yl methacrylate ≥98.0% (by GC) stabilized

Supplier: TCI America

Synonyms: GBLMA, 2-Methylacrylic acid 2-Oxotetrahydrofuran-3-yl ester

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

O0535-25G O0535-5G
TCO0535-25GEA 416 USD
TCO0535-25G TCO0535-5G
2-Oxotetrahydrofuran-3-yl methacrylate ≥98.0% (by GC) stabilized
2-Oxotetrahydrofuran-3-yl methacrylate
Formula: C₈H₁₀O₄
MW: 170.16 g/mol
MDL Number: MFCD19689400
CAS Number: 195000-66-9

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR