1-(4-Morpholinophenyl)-1-phenylprop-2-yn-1-ol ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M3184-1G M3184-5G
TCM3184-1GEA 62 USD
TCM3184-1G TCM3184-5G
1-(4-Morpholinophenyl)-1-phenylprop-2-yn-1-ol ≥98.0% (by HPLC, titration analysis)
1-(4-Morpholinophenyl)-1-phenylprop-2-yn-1-ol
Formula: C₁₉H₁₉NO₂
MW: 293.37 g/mol
CAS Number: 194940-93-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 158.0 to 162.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR