Dinaphtho[2,1-b:1',2'-d]furan ≥98.0% (by GC)

Supplier: TCI America

D5565-1G D5565-5G
TCD5565-1GEA 153 USD
TCD5565-1G TCD5565-5G
Dinaphtho[2,1-b:1',2'-d]furan ≥98.0% (by GC)
Dinaphtho[2,1-b:1',2'-d]furan
Formula: C₂₀H₁₂O
MW: 268.32 g/mol
MDL Number: MFCD30833908
CAS Number: 194-63-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 157.0 to 161.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR