4,6-Diphenyl-1,3,5-triazin-2(1H)-one ≥98.0% (by total nitrogen basis)

Supplier: TCI America

D5548-1G D5548-5G
TCD5548-1GEA 41 USD
TCD5548-1G TCD5548-5G
4,6-Diphenyl-1,3,5-triazin-2(1H)-one ≥98.0% (by total nitrogen basis)
4,6-Diphenyl-1,3,5-triazin-2(1H)-one
Formula: C₁₅H₁₁N₃O
MW: 249.27 g/mol
MDL Number: MFCD00234864
CAS Number: 1917-44-8
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR