1-Isopropylpyrazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1-Isopropyl-1H-pyrazole

I0999-1ML I0999-5ML
TCI0999-1MLEA 58 USD
TCI0999-1ML TCI0999-5ML
1-Isopropylpyrazole ≥98.0% (by GC, titration analysis)
1-Isopropylpyrazole
Formula: C₆H₁₀N₂
MW: 110.16 g/mol
Boiling Pt: 142 °C
MDL Number: MFCD07468920
CAS Number: 18952-87-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR