1,4-Diaminobutane dihydrobromide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Putrescine dihydrobromide, Tetramethylenediamine dihydrobromide, 1,4-Butanediamine dihydrobromide
D5685-1G
D5685-5G
TCD5685-1GEA
45
USD
TCD5685-1G
TCD5685-5G
1,4-Diaminobutane dihydrobromide ≥98.0% (by titrimetric analysis)
1,4-Diaminobutane dihydrobromide
Formula:
C₄H₁₂N₂·₂HBr MW: 249.98 g/mol |
CAS Number:
18773-04-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...