8-Pentadecanamine ≥96.0% (by GC, titration analysis)

Supplier: TCI America
Danger

P2543-1ML P2543-5ML
TCP2543-1MLEA 245 USD
TCP2543-1ML TCP2543-5ML
8-Pentadecanamine ≥96.0% (by GC, titration analysis)
8-Pentadecanamine
Formula: C₁₅H₃₃N
Boiling Pt: 117 °C
MDL Number: MFCD00087197
CAS Number: 18618-64-9
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR