Triethoxy(p-tolyl)silane ≥95.0% (by GC)

Supplier: TCI America
Warning

T3750-1G T3750-5G
TCT3750-1GEA 42 USD
TCT3750-1G TCT3750-5G
Triethoxy(p-tolyl)silane ≥95.0% (by GC)
Triethoxy(p-tolyl)silane
Formula: C₁₃H₂₂O₃Si
MW: 254.40 g/mol
Boiling Pt: 106 °C
MDL Number: MFCD04974099
CAS Number: 18412-57-2

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR