L-778123 dihydrochloride ≥98.0% (by HPLC)

Supplier: TCI America
Warning

L0337-25MG L0337-5MG
TCL0337-25MGEA 755 USD
TCL0337-25MG TCL0337-5MG
L-778123 dihydrochloride ≥98.0% (by HPLC)
L-778123 dihydrochloride
Formula: C₂₂H₂₀ClN₅O·₂HCl
Storage Temperature: Refrigerator
CAS Number: 183499-56-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 13.20 to 15.40 %
Chloride 14.1 to 15.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR