Formamidine Thiocyanate ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Formamidinium Thiocyanate

F1153-1G F1153-5G
TCF1153-1GEA 99 USD
TCF1153-1G TCF1153-5G
Formamidine Thiocyanate ≥98.0% (by total nitrogen basis)
Formamidine Thiocyanate
Formula: C₂H₅N₃S
MW: 103.14 g/mol
CAS Number: 1821033-48-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR