1,1,5,5-Tetramethyl-3,3-diphenyltrisiloxane ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Bis(dimethylsiloxy)diphenylsilane

T3832-100G T3832-25G
TCT3832-100GEA 167 USD
TCT3832-100G TCT3832-25G
1,1,5,5-Tetramethyl-3,3-diphenyltrisiloxane ≥98.0% (by GC)
1,1,5,5-Tetramethyl-3,3-diphenyltrisiloxane
Formula: C₁₆H₂₄O₂Si₃
MW: 332.62 g/mol
Boiling Pt: 83 °C
MDL Number: MFCD00411646
CAS Number: 17875-55-7
EINECS: 241-828-4

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR