4-Ethynyl-N,N-dimethylaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Dimethylaminophenylacetylene, 4-Ethynyl-N,N-dimethylbenzenamine, 4-Dimethylaminophenyl acetylene

E1021-1G E1021-5G
TCE1021-1GEA 135 USD
TCE1021-1G TCE1021-5G
4-Ethynyl-N,N-dimethylaniline ≥98.0% (by GC, titration analysis)
4-Ethynyl-N,N-dimethylaniline
Formula: C₁₀H₁₁N
MW: 145.21 g/mol
MDL Number: MFCD00168816
CAS Number: 17573-94-3

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 49.0 to 53.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR