Tizoxanide ≥95.0% (by HPLC)

Supplier: TCI America
Warning

T3862-100MG T3862-25MG
TCT3862-100MGEA 306 USD
TCT3862-100MG TCT3862-25MG
Tizoxanide ≥95.0% (by HPLC)
Tizoxanide
Formula: C₁₀H₇N₃O₄S
Melting Pt: 254 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07484970
CAS Number: 173903-47-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 15.10 to 16.60 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR