Trimethylsilyl Propionate ≥95.0% (by GC)

Supplier: TCI America
Danger

Synonyms: Propionic acid trimethylsilyl ester

T3552-1ML T3552-5ML
TCT3552-1MLEA 71 USD
TCT3552-1ML TCT3552-5ML
Trimethylsilyl Propionate ≥95.0% (by GC)
Trimethylsilyl Propionate
Formula: C₆H₁₄O₂Si
Boiling Pt: 122 °C
MDL Number: MFCD01632555
CAS Number: 16844-98-7
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR