2,2,5-Trimethyl-1,3-dioxane-5-carboxylic acid ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Warning

T3839-25G T3839-5G
TCT3839-25GEA 212 USD
TCT3839-25G TCT3839-5G
2,2,5-Trimethyl-1,3-dioxane-5-carboxylic acid ≥95.0% (by titrimetric analysis)
2,2,5-Trimethyl-1,3-dioxane-5-carboxylic acid
Formula: C₈H₁₄O₄
MDL Number: MFCD08272187
CAS Number: 16837-14-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 93.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 119.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR