Lanthanum(III) bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Bis(trifluoromethanesulphonyl)imide Lanthanum(III) salt, Lanthanum(III) triflimide

L0294-1G L0294-5G
TCL0294-1GEA 137 USD
TCL0294-1G TCL0294-5G
Lanthanum(III) bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)
Lanthanum(III) bis(trifluoromethanesulfonyl)imide
Formula: C₆F₁₈LaN₃O₁₂S₆
MW: 979.31 g/mol
CAS Number: 168106-26-1
UN: 1759
ADR: 8 II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR