1-(2,6-Diethylphenyl)-2,2,4-trimethyl-4-phenyl-3,4-dihydro-2H-pyrrol-1-ium tetrafluoroborate ≥98.0% (by HPLC)

Supplier: TCI America
Danger

D5655-1G D5655-5G
TCD5655-1GEA 258 USD
TCD5655-1G TCD5655-5G
1-(2,6-Diethylphenyl)-2,2,4-trimethyl-4-phenyl-3,4-dihydro-2H-pyrrol-1-ium tetrafluoroborate ≥98.0% (by HPLC)
1-(2,6-Diethylphenyl)-2,2,4-trimethyl-4-phenyl-3,4-dihydro-2H-pyrrol-1-ium tetrafluoroborate
Formula: C₂₃H₃₀BF₄N
MW: 407.30 g/mol
CAS Number: 1671098-46-6
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR