BDP FL ≥97.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 4,4-Difluoro-5,7-dimethyl-4-bora-3a,4a-diaza-s-indacene-3-propionic acid

D5554-25MG D5554-5MG
TCD5554-25MGEA 743 USD
TCD5554-25MG TCD5554-5MG
BDP FL ≥97.0% (by HPLC)
BDP FL
Formula: C₁₄H₁₅BF₂N₂O₂
MW: 292.09 g/mol
Melting Pt: 196 °C
MDL Number: MFCD00467384
CAS Number: 165599-63-3

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Nitrogen) 9.10 to 10.10 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR