2-Chloro-1,3-bis(2,6-diisopropylphenyl)-1H-imidazol-3-ium chloride cesium fluoride complex

Supplier: TCI America
Danger

P2420-1G P2420-5G
TCP2420-1GEA 293 USD
TCP2420-1G TCP2420-5G
2-Chloro-1,3-bis(2,6-diisopropylphenyl)-1H-imidazol-3-ium chloride cesium fluoride complex
2-Chloro-1,3-bis(2,6-diisopropylphenyl)-1H-imidazol-3-ium chloride cesium fluoride complex
Formula: C₂₇H₃₆Cl₂CsFN₂
MDL Number: MFCD29472387
CAS Number: 1648825-53-9
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Assay 30.0 to 32.0w% [as 2-Chloro-1,3-bis (2,6-diisopropylphenyl)-1H-imidazolium Chloride]


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR