5,7-Di-tert-butyl-3-(3,4-dimethylphenyl)benzofuran-2(3H)-one ≥98.0% (by GC)

Supplier: TCI America

D5640-25G D5640-5G
TCD5640-25GEA 115 USD
TCD5640-25G TCD5640-5G
5,7-Di-tert-butyl-3-(3,4-dimethylphenyl)benzofuran-2(3H)-one ≥98.0% (by GC)
5,7-Di-tert-butyl-3-(3,4-dimethylphenyl)benzofuran-2(3H)-one
Formula: C₂₄H₃₀O₂
MDL Number: MFCD30729989
CAS Number: 164391-52-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 131.0 to 135.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR